Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicer Mouse anti-Human, Clone: OTI1G8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526507

Catalog No. PIMA526507


Add to cart

Description

Description

DICER1 is a ribonuclease that has been extensively shown to be related to cancer. Small interfering RNAs are generated through DICER, and seem to play important roles in mitosis in normal cells and oncogenesis in mutated cells. DICER1 mutations are associated with blastoma, breast cancer, adenocarcinoma, and pancreatitis. In vision and hearing, DICER1 has been implicated in serious microRNA induced malformations of critical sensory cells. DICER1 is important in myelination and other nervous system protections. Finally, DICER1 is related to cardiovascular diseases, since microRNAs are aberrantly expressed in some pathological conditions.
Specifications

Specifications

Dicer
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UPY3
DCR1, Dicer, HERNA, MNG1, Dicer1, Dcr-1 homolog, K12H4.8-LIKE, dicer 1, double-stranded RNA-specific endoribonuclease, endoribonuclease Dicer, helicase MOI, helicase with RNAse motif, helicase-moi
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23405
Immunohistochemistry (Paraffin)
OTI1G8
Unconjugated
DICER1
Liquid
DICER1
Human recombit protein fragment corresponding to amino acids 1666-1922 of DICER1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit