Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%

Manufacturer: Alfa Aesar™ H2689506

Quantity 5g

 View more versions of this product

Catalog No. AAH2689506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67292-34-6
    Molecular Formula C34H28Cl2FeNiP2
    Formula Weight 684
    MDL Number MFCD00270284
    Synonym [1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
    Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)

    Specifications

    Formula Weight 684
    MDL Number MFCD00270284
    Melting Point 285°C
    Quantity 5g
    Molecular Formula C34H28Cl2FeNiP2
    CAS 67292-34-6
    Synonym [1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H334-H351-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
    Assay 99%

    Description

    Applications
    diphosphine 1,1 ft.-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents

    GHS H Statement
    H334-H351-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit