Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%

Manufacturer:  Alfa Aesar™ H2689514

Quantity 25g

 View more versions of this product

Catalog No. AAH2689514


  Check Availability
Add to cart

Specifications

Description

Applications
diphosphine 1,1 ft.-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents
Description & Specifications

Chemical Identifiers

67292-34-6
684
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
C34H28Cl2FeNiP2
MFCD00270284
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)

Specifications

67292-34-6
684
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
25g
99%
C34H28Cl2FeNiP2
MFCD00270284
285°C
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit