Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with acetone, Pd 13.0-15.0%

Coupling reactions, carbonylation

Manufacturer: Alfa Aesar™ 4497214

Quantity 25g

 View more versions of this product

Catalog No. AA4497214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 851232-71-8
    Molecular Formula C34H28Cl2FeP2Pd·(CH3)2CO
    Formula Weight 787.81
    MDL Number MFCD00792899
    Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride; PdCl2(dppf).(CH3)2CO
    Chemical Name or Material Dichloro[1,1'-bis (diphenylphosphino)ferrocene]palladium (II), Complex with acetone

    Specifications

    Formula Weight 787.81
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Molecular Formula C34H28Cl2FeP2Pd·(CH3)2CO
    CAS 851232-71-8
    Health Hazard 1 Warning
    Quantity 25g
    MDL Number MFCD00792899
    Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride; PdCl2(dppf).(CH3)2CO
    Color Orange-red
    Melting Point 280°C
    Concentration Pd 13.0 to 15.0%
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Dichloro[1,1'-bis (diphenylphosphino)ferrocene]palladium (II), Complex with acetone

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit