Alfa Aesar™ Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II), Pd 18.5%

Manufacturer: Alfa Aesar™ 1877909

Quantity 10g

 View more versions of this product

Catalog No. AA1877909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19978-61-1
    Linear Formula PdCl2[(C6H5)2PCH2CH2P(C6H5)2]
    Formula Weight 575.75
    MDL Number MFCD00015702
    Synonym [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II); [1,2-Bis(diphenylphosphino)ethane]palladium(II) chloride
    Chemical Name or Material Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)

    Specifications

    Formula Weight 575.75
    Physical Form Powder
    Linear Formula PdCl2[(C6H5)2PCH2CH2P(C6H5)2]
    MDL Number MFCD00015702
    CAS 19978-61-1
    Melting Point >360°C
    Synonym [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II); [1,2-Bis(diphenylphosphino)ethane]palladium(II) chloride
    Sensitivity Hygroscopic
    Quantity 10g
    Color Dark yellow
    Concentration Pd 18.5%
    Chemical Name or Material Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit