Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro(1,5-cyclooctadiene)platinum(II), 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
12080-32-9
C8H12Cl2Pt
374.18
MFCD00012413
VVAOPCKKNIUEEU-PHFPKPIQSA-L
dichloro 1,5-cyclooctadiene platinum ii,1,5-cyclooctadienedichloroplatinum,dichloro 1,5-cyclooctadiene platinum,platinum, dichloro 1,5-cyclooctadiene,1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride,platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene,1,5-cyclooctadieneplatinum ii dichloride,dichloro 1,5-cyclooctadiene platinum ii 250mg,dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum,platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
6436378
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
C1CC=CCCC=C1.[Cl-].[Cl-].[Pt+2]
12080-32-9
C8H12Cl2Pt
374.18
MFCD00012413
VVAOPCKKNIUEEU-PHFPKPIQSA-L
dichloro 1,5-cyclooctadiene platinum ii,1,5-cyclooctadienedichloroplatinum,dichloro 1,5-cyclooctadiene platinum,platinum, dichloro 1,5-cyclooctadiene,1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride,platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene,1,5-cyclooctadieneplatinum ii dichloride,dichloro 1,5-cyclooctadiene platinum ii 250mg,dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum,platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
6436378
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
C1CC=CCCC=C1.[Cl-].[Cl-].[Pt+2]
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 285.0°C
Color White to Yellow
Assay Percent Range 97.5% min. (ex Pt)
Solubility Information Solubility in water: insoluble in water
Formula Weight 374.18
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Dichloro(1, 5-cyclooctadiene)platinum(II)
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 235-144-5