Alfa Aesar™ Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%

Manufacturer: Alfa Aesar™ 4497106

Quantity 5g

 View more versions of this product

Catalog No. AA4497106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 29964-62-3
    Molecular Formula C28H28Cl2P2Pd
    Formula Weight 603.8
    MDL Number MFCD02093437
    Chemical Name or Material Dichloro[bis (1,4-diphenylphosphino)butane]palladium (II)

    Specifications

    Physical Form Lumps
    Formula Weight 603.8
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Concentration Pd 17.6%
    MDL Number MFCD02093437
    CAS 29964-62-3
    Health Hazard 1 Warning
    Quantity 5g
    Molecular Formula C28H28Cl2P2Pd
    Chemical Name or Material Dichloro[bis (1,4-diphenylphosphino)butane]palladium (II)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit