Alfa Aesar™ Dichloro(norbornadiene)platinum(II), Pt 54.4%

Manufacturer: Alfa Aesar™ 4403506

Quantity 5g

 View more versions of this product

Catalog No. AA4403506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12152-26-0
    Formula Weight 358.13
    MDL Number MFCD00213423
    Synonym PtCl2(nbd); (Bicyclo[2.2.1]hepta-2,5-diene)dichloroplatinum(II)
    Chemical Name or Material Dichloro(norbornadiene)platinum(II)

    Specifications

    Formula Weight 358.13
    MDL Number MFCD00213423
    Solubility Information Soluble in acetic acid
    Physical Form Crystalline
    CAS 12152-26-0
    Synonym PtCl2(nbd); (Bicyclo[2.2.1]hepta-2,5-diene)dichloroplatinum(II)
    Storage Note 1 2° to 8°C
    Concentration Pt 54.4%
    Color Cream
    Melting Point ≈200°C (dec.)
    Quantity 5g
    Chemical Name or Material Dichloro(norbornadiene)platinum(II)

    2° to 8°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit