Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Dichloro-p-xylene, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
623-25-6
C8H8Cl2
175.06
MFCD00000920
ZZHIDJWUJRKHGX-UHFFFAOYSA-N
1,4-bis chloromethyl benzene,alpha,alpha'-dichloro-p-xylene,p-xylylene dichloride,p-xylylene chloride,benzene, 1,4-bis chloromethyl,p-bis chloromethyl benzene,benzene, p-bis chloromethyl,a,a'-dichloro-p-xylene,xylylene dichloride,ccris 1772
12171
1,4-bis(chloromethyl)benzene
C1=CC(=CC=C1CCl)CCl
623-25-6
C8H8Cl2
175.06
MFCD00000920
ZZHIDJWUJRKHGX-UHFFFAOYSA-N
1,4-bis chloromethyl benzene,alpha,alpha'-dichloro-p-xylene,p-xylylene dichloride,p-xylylene chloride,benzene, 1,4-bis chloromethyl,p-bis chloromethyl benzene,benzene, p-bis chloromethyl,a,a'-dichloro-p-xylene,xylylene dichloride,ccris 1772
12171
1,4-bis(chloromethyl)benzene
C1=CC(=CC=C1CCl)CCl
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 98.0°C to 102.0°C
Boiling Point 254.0°C
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H4(CH2Cl)2
Beilstein 05, 384
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in acetone and methylenechloride,50mg/mL methanol
Formula Weight 175.06
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material α, α'-Dichloro-p-xylene, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-782-7

RTECSNumber : ZE4100000

TSCA : TSCA