Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Dichloro-p-xylene, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
623-25-6
C8H8Cl2
175.052
MFCD00000920
ZZHIDJWUJRKHGX-UHFFFAOYSA-N
1,4-bis chloromethyl benzene,alpha,alpha'-dichloro-p-xylene,p-xylylene dichloride,p-xylylene chloride,benzene, 1,4-bis chloromethyl,p-bis chloromethyl benzene,benzene, p-bis chloromethyl,a,a'-dichloro-p-xylene,xylylene dichloride,ccris 1772
12171
1,4-bis(chloromethyl)benzene
C1=CC(=CC=C1CCl)CCl
623-25-6
C8H8Cl2
175.052
MFCD00000920
ZZHIDJWUJRKHGX-UHFFFAOYSA-N
1,4-bis chloromethyl benzene,alpha,alpha'-dichloro-p-xylene,p-xylylene dichloride,p-xylylene chloride,benzene, 1,4-bis chloromethyl,p-bis chloromethyl benzene,benzene, p-bis chloromethyl,a,a'-dichloro-p-xylene,xylylene dichloride,ccris 1772
12171
1,4-bis(chloromethyl)benzene
C1=CC(=CC=C1CCl)CCl
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Yellow
Quantity 500g
Melting Point 98.0°C to 102.0°C
Boiling Point 254.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H4(CH2Cl)2
Beilstein 05, 384
Formula Weight 175.06
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material α, α'-Dichloro-p-xylene, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-782-7

RTECSNumber : ZE4100000

TSCA : TSCA