Learn More
Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%, Thermo Scientific Chemicals
Description
Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer is used as a catalyst for amidation, reductive alkylation, hydrogenation, chiral hydrogenation reactions. It is also used as a catalyst for oxidative olefination reactions, C-C bond cleavage of secondary alcohols, oxidative annulation of pyridine and ortho C-H olefination of phenol derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Color | Red |
Odor | Odorless |
Quantity | 250 mg |
Solubility Information | Soluble in chloroform and acetone. Slightly soluble in tetrahydrofuran and methanol. Insoluble in water and diethylether |
Formula Weight | 618.08 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 625-704-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.