Alfa Aesar™ Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer

Manufacturer: Alfa Aesar™ 4634803

Quantity 1g

 View more versions of this product

Catalog No. AA4634803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 96503-27-4
    Formula Weight 307.21
    MDL Number MFCD00070449
    Synonym (RuCl2Cp*)n
    Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer

    Specifications

    Synonym (RuCl2Cp*)n
    Formula Weight 307.21
    Melting Point 45° to 47°C
    CAS 96503-27-4
    Color Red-brown
    Physical Form Crystalline Solid
    MDL Number MFCD00070449
    Storage Note 1 2° to 8°C
    Sensitivity Air & Moisture Sensitive
    Quantity 1g
    Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit