Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetic acid, 99+%, Thermo Scientific™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
79-43-6
C2H2Cl2O2
128.94
MFCD00004223
JXTHNDFMNIQAHM-UHFFFAOYSA-N
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
6597
CHEBI:36386
2,2-dichloroacetic acid
C(C(=O)O)(Cl)Cl
79-43-6
C2H2Cl2O2
128.94
MFCD00004223
JXTHNDFMNIQAHM-UHFFFAOYSA-N
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
6597
CHEBI:36386
2,2-dichloroacetic acid
C(C(=O)O)(Cl)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 1.5600g/mL
Color Brown
Melting Point 9.0°C to 11.0°C
Boiling Point 194.0°C
Flash Point >112°C
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula Cl2CHCO2H
Beilstein 02,202
Packaging Glass bottle
Fieser 16,115
Merck Index 15,3059
Refractive Index 1.4653 to 1.4673
Quantity 1L
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,ether and ethanol,soluble in acetone
Specific Gravity 1.56
Formula Weight 128.94
Physical Form Crystals or Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Dichloroacetic acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Suspected of causing cancer.
May damage fertility or the unborn child.
May cause harm to breast-fed children.
May cause damage to or

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 201-207-

RUO – Research Use Only