Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetic anhydride, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
4124-30-5
C4H2Cl4O3
239.87
MFCD00000842
RQHMQURGSQBBJY-UHFFFAOYSA-N
dichloroacetic anhydride,dichloroacetic acid anhydride,acetic acid, dichloro-, anhydride,unii-mb3ogr64in,anhydrid kyseliny dichloroctove,2,2'-dichloroacetic anhydride,dichloroacetetic anhydride,mb3ogr64in,anhydrid kyseliny dichloroctove czech,acetic acid, 2,2-dichloro-, 1,1'-anhydride
77772
(2,2-dichloroacetyl) 2,2-dichloroacetate
C(C(=O)OC(=O)C(Cl)Cl)(Cl)Cl
4124-30-5
C4H2Cl4O3
239.87
MFCD00000842
RQHMQURGSQBBJY-UHFFFAOYSA-N
dichloroacetic anhydride,dichloroacetic acid anhydride,acetic acid, dichloro-, anhydride,unii-mb3ogr64in,anhydrid kyseliny dichloroctove,2,2'-dichloroacetic anhydride,dichloroacetetic anhydride,mb3ogr64in,anhydrid kyseliny dichloroctove czech,acetic acid, 2,2-dichloro-, 1,1'-anhydride
77772
(2,2-dichloroacetyl) 2,2-dichloroacetate
C(C(=O)OC(=O)C(Cl)Cl)(Cl)Cl
Color Colorless
Quantity 10g
Flash Point >112°C
Density 1.5700g/mL
Packaging Glass bottle
Boiling Point 214.0°C to 216.0°C
Melting Point 29.0°C
Specific Gravity 1.57
Refractive Index 1.4825 to 1.4845
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula (Cl2CHCO)2O
Beilstein 02, 204
Solubility Information Solubility in water: decomposes in water. Other solubilities: decomposes in alcohol
Formula Weight 239.87
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Dichloroacetic anhydride, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 223-924-8

RTECSNumber : AG6300000

TSCA : TSCA