Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Dichloroacetyl chloride, 97%
Click to view available options
Quantity:
100g
500g
Specifications
Flash Point | None |
Density | 1.532 |
Refractive Index | 1.462 |
Sensitivity | Moisture sensitive |
Boiling Point | 107°C to 108°C |
Quantity | 500g |
UN Number | UN1765 |
Merck Index | 14,3053 |
Formula Weight | 147.39 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H311-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETYL CHLORIDE
EINECSNumber : 201-199-9
RTECSNumber : AO6650000
TSCA : Yes
Recommended Storage : Ambient temperatures