Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II), 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
1092372-91-2
C20H48Cl2N2P2Ru
550.54
MFCD07782003
FUALRLYVUZSWKT-UHFFFAOYSA-L
dichlorobis 2-di-tert-butylphosphino ethylamine ruthenium ii,dichlorobis 2-di-t-butylphosphino ethylamine ruthenium ii
71310624
2-ditert-butylphosphanylethanamine;dichlororuthenium
CC(C)(C)P(CCN)C(C)(C)C.CC(C)(C)P(CCN)C(C)(C)C.Cl[Ru]Cl
1092372-91-2
C20H48Cl2N2P2Ru
550.54
MFCD07782003
FUALRLYVUZSWKT-UHFFFAOYSA-L
dichlorobis 2-di-tert-butylphosphino ethylamine ruthenium ii,dichlorobis 2-di-t-butylphosphino ethylamine ruthenium ii
71310624
2-ditert-butylphosphanylethanamine;dichlororuthenium
CC(C)(C)P(CCN)C(C)(C)C.CC(C)(C)P(CCN)C(C)(C)C.Cl[Ru]Cl
Melting Point 247°C to 263°C
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Color Brown to Yellow
Elemental Analysis 41.9 to 45.4% C, 4.9 to 5.3% N
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure
Assay Percent Range 97%
Formula Weight 550.54
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning