Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorobis(2-(diisopropylphosphino)ethylamine)ruthenium(II), 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
1092372-90-1
C16H40Cl2N2P2Ru
494.428
MFCD07782005
HINVYXXIKGEKJI-UHFFFAOYSA-L
dichlorobis 2-diisopropylphosphino-ethylamine ruthenium ii,dichlorobis 2-di-i-propylphosphino ethylamine ruthenium ii,2-di propan-2-yl phosphanyl ethan-1-amine-dichlororuthenium 2/1
71310888
dichlororuthenium;2-di(propan-2-yl)phosphanylethanamine
CC(C)P(CCN)C(C)C.CC(C)P(CCN)C(C)C.Cl[Ru]Cl
1092372-90-1
C16H40Cl2N2P2Ru
494.428
MFCD07782005
HINVYXXIKGEKJI-UHFFFAOYSA-L
dichlorobis 2-diisopropylphosphino-ethylamine ruthenium ii,dichlorobis 2-di-i-propylphosphino ethylamine ruthenium ii,2-di propan-2-yl phosphanyl ethan-1-amine-dichlororuthenium 2/1
71310888
dichlororuthenium;2-di(propan-2-yl)phosphanylethanamine
CC(C)P(CCN)C(C)C.CC(C)P(CCN)C(C)C.Cl[Ru]Cl
Infrared Spectrum Authentic
Elemental Analysis 37.3 to 40.4% C
Quantity 250mg
Color Orange to Brown
NMR Consistent with structure
Assay Percent Range 97%
Formula Weight 494.43
Physical Form Crystalline Powder
Chemical Name or Material Dichlorobis(2-(diisopropylphosphino)ethylamine)ruthenium(II)

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.