Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II), 97%, Thermo Scientific™

Catalog Number AC435272500
Click to view available options
Quantity:
250 mg
1 g
911128-25-1
C30H36Cl2N2P2Ru
658.55
MFCD15144861
MLZFSEGMJUGESY-UHFFFAOYSA-L
71310857
dichlororuthenium;3-diphenylphosphanylpropan-1-amine
C1=CC=C(C=C1)P(CCCN)C2=CC=CC=C2.C1=CC=C(C=C1)P(CCCN)C2=CC=CC=C2.Cl[Ru]Cl
911128-25-1
C30H36Cl2N2P2Ru
658.55
MFCD15144861
MLZFSEGMJUGESY-UHFFFAOYSA-L
71310857
dichlororuthenium;3-diphenylphosphanylpropan-1-amine
C1=CC=C(C=C1)P(CCCN)C2=CC=CC=C2.C1=CC=C(C=C1)P(CCCN)C2=CC=CC=C2.Cl[Ru]Cl
Color Yellow to Brown
Elemental Analysis 52.5 to 56.9% C, 4.1 to 4.4% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 658.55
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit