Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorodimethylsilane, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
Packaging:
Glass bottle
75-78-5
C2H6Cl2Si
129.06
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
6398
dichloro(dimethyl)silane
C[Si](C)(Cl)Cl
75-78-5
C2H6Cl2Si
129.06
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
6398
dichloro(dimethyl)silane
C[Si](C)(Cl)Cl
Viscosity 0.6 mPa.s (25°C)
Specific Gravity 1.06
Refractive Index 1.4030 to 1.4050
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5mL
Flash Point −9°C
Melting Point -76.0°C
Boiling Point 70.0°C
Density 1.0600g/mL
Color Colorless to Yellow
Assay Percent Range 99% min. (GC)
Linear Formula (CH3)2SiCl2
Fieser 03,114; 04,183; 17,113
Solubility Information Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
Formula Weight 129.06
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Dichlorodimethylsilane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 200-901-