Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorofluorescein, 0.2% In 70% Isopropanol, Reagents

Catalog No. p-200022059
Click to view available options
Quantity:
100 mL
500 mL

Dichlorofluorescein, 0.2% In 70% Isopropanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0, 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym 2',7'-Dichloro-3,6-fluorandiol, 2,7-Dichlorofluorescein, 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Boiling Point 82°C
Percent Purity 0.2%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.86
Color Yellow/Green
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Dichlorofluorescein, 0.2% In 70% Isopropanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.