Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Dichloroisobutylmethylsilane, 97%
Click to view available options
Quantity:
5g
25g
100g
Specifications
UN Number | UN2987 |
Formula Weight | 171.14 |
Percent Purity | 97% |
Refractive Index | 1.4295 |
Sensitivity | Moisture sensitive |
Boiling Point | 130°C |
Quantity | 5g |
Flash Point | 64°C (147°F) |
Chemical Name or Material | Dichloroisobutylmethylsilane |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber : 241-942-4
TSCA : No
Recommended Storage : Ambient temperatures