Alfa Aesar™ Dichloroisocyanuric acid sodium salt, 97% (dry wt.), water <3%

Manufacturer: Alfa Aesar™ B2350436

Quantity 500g

 View more versions of this product

Catalog No. AAB2350436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2893-78-9
    Molecular Formula C3Cl2N3NaO3
    Formula Weight 219.95
    MDL Number MFCD00006036
    UN Number UN2465
    Beilstein 4056155
    Synonym 1,3-Dichloro-6-hydroxy-1,3,5-triazine-2,4-dione sodium salt; Sodium dichloroisocyanurate
    Chemical Name or Material Dichloroisocyanuric acid sodium salt

    Specifications

    Formula Weight 219.95
    CAS 2893-78-9
    UN Number UN2465
    Melting Point ≈250°C (dec.)
    Synonym 1,3-Dichloro-6-hydroxy-1,3,5-triazine-2,4-dione sodium salt; Sodium dichloroisocyanurate
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number XZ1900000
    Quantity 500g
    TSCA TSCA
    Odor Chlorine-like
    Molecular Formula C3Cl2N3NaO3
    MDL Number MFCD00006036
    Beilstein 4056155
    Health Hazard 2 GHS H Statement
    H272-H302-H319-H335
    May intensify fire; oxidizer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: DICHLOROISOCYANURIC ACID, SALTS
    Chemical Name or Material Dichloroisocyanuric acid sodium salt
    Assay 97%, (dry wt.), water <3%

    Description

    Applications
    Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters, synthesis of antimicrobial polyurethane additives to control biofilm. It is used as a emergency disinfectant in drinking water, widely used cleansing agent. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.

    Solubility
    Soluble in water.

    Notes
    Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

    GHS H Statement
    H272-H302-H319-H335
    May intensify fire; oxidizer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit