Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dichloroisocyanuric acid sodium salt, 97% (dry wt.), water <3%

Click to view available options
Quantity:
25g
100g
500g
2893-78-9
C3HCl2N3NaO3
220.949
MFCD00006036
UNWRHVZXVVTASG-UHFFFAOYSA-N
sodium dichlorocyanuric acid,1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
86657659
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]
2893-78-9
C3HCl2N3NaO3
220.949
MFCD00006036
UNWRHVZXVVTASG-UHFFFAOYSA-N
sodium dichlorocyanuric acid,1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
86657659
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]
Applications
Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters, synthesis of antimicrobial polyurethane additives to control biofilm. It is used as a emergency disinfectant in drinking water, widely used cleansing agent. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Melting Point ~250°C (decomposition)
Quantity 100g
Odor Chlorine-like
UN Number UN2465
Beilstein 4056155
Formula Weight 219.95
Percent Purity 97%
Chemical Name or Material Dichloroisocyanuric acid sodium salt
Assay (dry wt.), water <3%

GHS H Statement
H272-H302-H319-H335
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: DICHLOROISOCYANURIC ACID, SALTS

RTECSNumber : XZ1900000

TSCA : TSCA