Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroisocyanuric acid sodium salt, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
2893-78-9
C3HCl2N3NaO3
220.949
MFCD00006036
UNWRHVZXVVTASG-UHFFFAOYSA-N
sodium dichlorocyanuric acid,1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
86657659
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]
2893-78-9
C3HCl2N3NaO3
220.949
MFCD00006036
UNWRHVZXVVTASG-UHFFFAOYSA-N
sodium dichlorocyanuric acid,1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
86657659
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger

Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger

C-X bond formation reagent, Oxidation reagent
Quantity 25g
Color White
Packaging Plastic bottle
Water 5% max.
pH 6 to 7 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 219.95
Physical Form Crystals or Granules
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material Dichloroisocyanuric acid sodium salt, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 220-767-7

RTECSNumber : XZ1900000

TSCA : TSCA