Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroisocyanuric acid sodium salt monohydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
52671-45-1
C3Cl2N3NaO3·H2O
237.96
FIHQYHSNTSJNGG-UHFFFAOYSA-N
Sodium dichloroisocyanurate dihydrate
129893375
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium;dihydrate
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.O.O.[Na]
52671-45-1
C3Cl2N3NaO3·H2O
237.96
FIHQYHSNTSJNGG-UHFFFAOYSA-N
Sodium dichloroisocyanurate dihydrate
129893375
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium;dihydrate
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.O.O.[Na]
Oxidation reagent
Assay Percent Range 96.0 to 104% (on dry substance) (Iodometry)
Solubility Information Solubility in water: ca 250g/L (25°C)
Formula Weight 237.96
Physical Form Crystals or Granules
Percent Purity 97%
Free Chloride 0.5% max.
Infrared Spectrum Authentic
Loss on Drying 15% max.
Packaging Glass bottle
Solubility Solubility in water: ca 250g/L (25°C).
Color White
Melting Point 240.0°C to 250.0°C
Quantity 50g
Chemical Name or Material Dichloroisocyanuric acid sodium salt monohydrate
Show More Show Less

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates t

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger