Alfa Aesar™ Dichloromalononitrile, 96%

Manufacturer: Alfa Aesar™ B2177906

Quantity 5g

 View more versions of this product

Catalog No. AAB2177906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13063-43-9
    Molecular Formula C3Cl2N2
    Formula Weight 134.95
    MDL Number MFCD00001841
    UN Number UN3276
    Beilstein 906848
    Chemical Name or Material Dichloromalononitrile

    Specifications

    Formula Weight 134.95
    Odor Pungent
    MDL Number MFCD00001841
    UN Number UN3276
    Beilstein 906848
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Flash Point >110°C (230°F)
    Boiling Point 96° to 97°C
    Quantity 5g
    Molecular Formula C3Cl2N2
    CAS 13063-43-9
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    RTECS Number OO3740000
    Chemical Name or Material Dichloromalononitrile
    Assay 96%

    GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit