Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.5%, for analysis, stabilized with ethanol, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
75-09-2,64-17-5
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2,64-17-5
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
Specific Gravity 1.325
Viscosity 0.43 mPa.s (20°C)
Acidity 0.0005 meq/g max.
Assay Percent Range 99.4% min. (GC)
Linear Formula CH2Cl2
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and dmf
Formula Weight 84.93
Physical Form Liquid
Percent Purity 99.5%
Grade Analysis
Density 1.3250g/mL
Packaging Glass bottle
Refractive Index 1.4230 to 1.4250
Residue after Evaporation 0.0005% max.
Water 0.02% max. (K.F.)
Boiling Point 39.0°C to 40.0°C
Color Undesignated
Melting Point -97.0°C
Quantity 2.5L
Chemical Name or Material Dichloromethane
Show More Show Less

GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
Do not handle until all safety precautions have been read and understood.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: W

GHS Signal Word: Warning

EINECSNumber : 200-838-9