Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.8%, for HPLC, stabilized with methanol, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
75-09-2,67-56-1
CH2Cl2
84.927
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2,67-56-1
CH2Cl2
84.927
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
for HPLC
Absorbance (1 cm cell vs HPLC−grade water),0.005 max. at 260nm,0.005 max. at 270nm,0.005 max. at 280nm,0.02 max. at 250nm,0.2 max. at 240nm,1 max. at 233nm
Absorbance Conditions (1 cm cell vs HPLC-grade water)
Acidity 0.0001 meq/g max.
Assay Percent Range 99.80%
Linear Formula CH2Cl2
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Formula Weight 84.93
Physical Form Liquid
Percent Purity 99.8%
Grade HPLC
Density 1.3250g/mL
Filtered to 0.2 micrometer Yes
Free Chlorine 0.0005% max.
Packaging Glass bottle
Residue after Evaporation 0.0002% max.
Solubility Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Specific Gravity 1.325
Stabilizer approx. 0.4% methanol
Water 0.02% max.
Viscosity 0.43 mPa.s (20°C)
Boiling Point 39.0°C to 40.0°C
Color Undesignated
Melting Point -97.0°C
Quantity 2.5L
Chemical Name or Material Dichloromethane, Stabilized
Show More Show Less

GHS P Statement
Suspected of causing cancer.

GHS P Statement
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-838-9

RTECSNumber : PA8050000

TSCA : TSCA