Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, ACROS Organics™

Click to view available options
Quantity:
2.5L
10L
Packaging:
Glass bottle
Metal drum
75-09-2,513-35-9
CH2Cl2
84.927
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2,513-35-9
CH2Cl2
84.927
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
For peptide synthesis
Assay Percent Range 99.9%
Linear Formula CH2Cl2
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Physical Form Liquid
Percent Purity 99.9%
Density 1.3250g/mL
Free Amine 0.001% max.
Non_volatile Matter 0.0005% max.
Packaging Glass bottle
Solubility Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Specific Gravity 1.325
Stabilizer approx. 0.002% amylene
Water 0.005% max.
Viscosity 0.43 mPa.s (20°C)
Boiling Point 39.0°C to 40.0°C
Color Undesignated
Melting Point -97.0°C
Quantity 2.5L
Chemical Name or Material Dichloromethane, Stabilized with Amylene
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.

GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

Warning