Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Metal drum
75-09-2,513-35-9
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2,513-35-9
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
For residue analysis
Absorbance 0.5 max. at 235nm,0.108 max. at 240nm,0.009 max. at 260nm
Acidity 0.001% max. (HCl)
Appearance Clear liquid
Assay Percent Range 99.9%
Linear Formula CH2Cl2
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.9%
Grade Residue Analysis
Density 1.3250g/mL
Packaging Glass Bottle
Residue after Evaporation 0.0005% max.
Solubility Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Specific Gravity 1.325
Stabilizer 0.003 to 0.006 w/w% amylene
Water 0.02% max. (Coulometric)
Viscosity 0.43 mPa.s (20°C)
Boiling Point 39.0°C to 40.0°C
Color Undesignated
Melting Point -97.0°C
Quantity 1L
Chemical Name or Material Dichloromethane
Show More Show Less

GHS H Statement
Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.

Warning