Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloromethane 99+%, stabilized with amylene, pure, ACROS Organics™
Click to view available options
Quantity:
1L
2.5L
25L
Specifications
Assay Percent Range | 99+% |
Linear Formula | CH2Cl2 |
Fieser | 01,676; 02,273; 04,337; 07,239; 13,106 |
Solubility Information | Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf |
Formula Weight | 84.93 |
Percent Purity | 99+% |
Grade | Extra Pure |
Density | 1.3250g/mL |
Packaging | Glass Bottle |
Solubility | Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf |
Show More |
Safety and Handling
EINECSNumber : 200-838-9