Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
75-09-2
75-09-2
CAS Max % 1
Percent Purity 0.995
Grade ACS
Free Halogens To Pass Test
Identification To Pass Test
Packaging Steel Can
Residue after Evaporation 2e-05
Titratable Acid 0.0003 meq/g
Water 0.0002
Quantity 20 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.