Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, Free halogens P.T., H2O 0.02%

Manufacturer:  Alfa Aesar 39116M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA39116M1


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

75-09-2
>99.5%
84.93
Dichloromethane, For analysis ACS
Liquid
CH2Cl2
MFCD00000881
ACS Grade

Specifications

Colorless liquid
605°C (1121°F)
Liquid
CH2Cl2
MFCD00000881
Clear colorless
Dichloromethane, For analysis ACS
75-09-2
Sweet
>99.5%
84.93
475 hPa (356mm Hg) at 20°C
4 x 1L
ACS Grade

Safety and Handling

SDS & Safety Documents

H351: Suspected of causing cancer.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 200-838-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit