Alfa Aesar™ Dichloromethane-d2, 100% (Isotopic)

Manufacturer: Alfa Aesar™ 42334SA

Quantity 2 each

 View more versions of this product

Catalog No. AA42334SA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1665-00-5
    Formula Weight 86.95
    MDL Number MFCD00000882
    UN Number UN1593
    Chemical Name or Material Dichloromethane-d2, Isotopic

    Specifications

    Formula Weight 86.95
    Packaging Prescored Ampules
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Quantity 2 each
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    Odor Sweet
    MDL Number MFCD00000882
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
    Sensitivity Moisture Sensitive
    Density 1.35g/mL
    CAS 1665-00-5
    UN Number UN1593
    Chemical Name or Material Dichloromethane-d2, Isotopic
    Assay 100%

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit