Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane-d2, for NMR, 100.0 atom % D, Acros Organics

Click to view available options
Quantity:
5g
1665-00-5
CH2Cl2
86.939
YMWUJEATGCHHMB-DICFDUPASA-N
dichloromethane-d2,methylene chloride-d2,dichloro 2h2 methane,methane-d2, dichloro,cd2cl2,deuterated dichloromethane,dichloro dideuterio methane,dideuteromethylene chloride,dichloromethane-d2, 99.9 atom % d,dichloromethane-d', 99.96 atom % d
160586
dichloro(dideuterio)methane
C(Cl)Cl
1665-00-5
CH2Cl2
86.939
YMWUJEATGCHHMB-DICFDUPASA-N
dichloromethane-d2,methylene chloride-d2,dichloro 2h2 methane,methane-d2, dichloro,cd2cl2,deuterated dichloromethane,dichloro dideuterio methane,dideuteromethylene chloride,dichloromethane-d2, 99.9 atom % d,dichloromethane-d', 99.96 atom % d
160586
dichloro(dideuterio)methane
C(Cl)Cl
Color Colorless
Deuterium (D) 99.95% min.
Density 1.3600g/mL
Water 0.01% max. (Coulometric)
Quantity 5g
Linear Formula CD2Cl2
Beilstein 01, IV, 39
Formula Weight 86.95
Physical Form Liquid
Grade NMR
Chemical Name or Material Dichloromethane-d2, 100.0 atom % D
Show More Show Less

GHS H Statement:
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning