Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, HPLC Grade, >99.7%, Alfa Aesar™

Manufacturer:  Alfa Aesar 22917AK

Quantity 250mL

 View more versions of this product

Catalog No. AA22917AK


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

75-09-2
>99.7%
84.93
Dichloromethane, Stable with amylene
Liquid
CH2Cl2
MFCD00000881
HPLC Grade

Specifications

Colorless
75-09-2
Liquid
CH2Cl2
84.93
475 hPa (356mm Hg) at 20°C
HPLC Grade
250mL
Colorless liquid
>99.7%
Sweet
MFCD00000881
Dichloromethane, Stable with amylene

Safety and Handling

SDS & Safety Documents

H351: Suspected of causing cancer.

DOTInformation : DOT Class: 6.1; Packing Group: III

EINECSNumber : 200-838-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit