Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
4 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Melting Point | -97.0°C |
Decomposition Information | >120°C |
Density | 1.3250g/mL |
Boiling Point | 39.0°C to 40.0°C |
Quantity | 1 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Residue after Evaporation | 2ppm max. |
Specific Gravity | 1.325 |
Linear Formula | CH2Cl2 |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician. |
EINECSNumber | 200-838-9 |
RTECSNumber | PA8050000 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction