Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Spectrophotometric Grade, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
75-09-2
75-09-2
CAS Max % 1
Appearance Clear
Percent Purity 0.995
Grade Spectrophotometric
Free Halogens To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Titratable Acid 0.0003 meq/g
Water 0.0002
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.