Alfa Aesar™ Dichloromethylphenylsilane, 98%

Manufacturer: Alfa Aesar™ B2399922

Quantity 100g

 View more versions of this product

Catalog No. AAB2399922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 149-74-6
    Molecular Formula C7H8Cl2Si
    Linear Formula C6H5Si(CH3)Cl2
    Formula Weight 191.13
    MDL Number MFCD00000490
    UN Number UN2437
    Beilstein 970975
    Synonym Methylphenyldichlorosilane; Phenylmethyldichlorosilane
    Chemical Name or Material Dichloromethylphenylsilane

    Specifications

    Formula Weight 191.13
    UN Number UN2437
    Flash Point 82°C (179°F)
    TSCA TSCA
    Odor Acrid
    Molecular Formula C7H8Cl2Si
    Linear Formula C6H5Si(CH3)Cl2
    CAS 149-74-6
    Synonym Methylphenyldichlorosilane; Phenylmethyldichlorosilane
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHYLPHENYLDICHLOROSILANE
    RTECS Number VV3530000
    MDL Number MFCD00000490
    Beilstein 970975
    Boiling Point 205° to 206°C
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Chemical Name or Material Dichloromethylphenylsilane
    Assay 98%

    Description

    Applications
    Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

    Solubility
    Reacts with water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P280-P305+P351+P338-P309-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit