Learn More
Dichloromethylphenylsilane, 98%, Thermo Scientific Chemicals
Description
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.
Specifications
Boiling Point | 205°C to 206°C |
Quantity | 500 g |
Linear Formula | C6H5Si(CH3)Cl2 |
UN Number | UN2437 |
Beilstein | 970975 |
Flash Point | 82°C (179°F) |
Solubility Information | Reacts with water. |
Formula Weight | 191.13 |
Percent Purity | 98% |
Odor | Acrid |
Show More |
Safety and Handling
Hazard Category | H227-H302-H314 |
Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
Precautionary Statement | P210-P260-P264b-P270-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHYLPHENYLDICHLOROSILANE |
EINECSNumber | 205-746-2 |
RTECSNumber | VV3530000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.