Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
75-54-7
CH4Cl2Si
MFCD00000494
Methyldichlorosilane
75-54-7
CH4Cl2Si
MFCD00000494
Methyldichlorosilane
Reduction reagent
Density 1.1000g/mL
Quantity 100mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.3970 to 1.4000 (20°C, 589nm)
Color Yellow to Brown
Assay Percent Range 97%
Linear Formula CH3SiHCl2
Beilstein 04, I, 581
Formula Weight 115.04
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Dichloromethylsilane, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if inhaled.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-877-1

RTECSNumber : VV3500000

TSCA : TSCA