Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorodimethylsilane, 99+%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
75-78-5
C2H6Cl2Si
129.06
MFCD00000491
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dimethyldichlorosilane,silane, dichlorodimethyl,dichloro dimethyl silane,inerton aw-dmcs,dichlorodimethylsilicon,dimethyl-dichlorsilan,inerton dw-dmc,repel-silan,dimethylsilane dichloride,dimethyl dichlorosilane
6398
dichloro(dimethyl)silane
C[Si](C)(Cl)Cl
75-78-5
C2H6Cl2Si
129.06
MFCD00000491
LIKFHECYJZWXFJ-UHFFFAOYSA-N
dimethyldichlorosilane,silane, dichlorodimethyl,dichloro dimethyl silane,inerton aw-dmcs,dichlorodimethylsilicon,dimethyl-dichlorsilan,inerton dw-dmc,repel-silan,dimethylsilane dichloride,dimethyl dichlorosilane
6398
dichloro(dimethyl)silane
C[Si](C)(Cl)Cl
Specific Gravity 1.06
Viscosity 0.6 mPa.s (25°C)
Boiling Point 70.0°C
Packaging AcroSeal™ Glass Bottle
Quantity 800mL
Infrared Spectrum Authentic
Melting Point -76.0°C
Flash Point −9°C
Color Colorless to Yellow
Refractive Index 1.4030 to 1.4050
Density 1.0600g/mL
Assay Percent Range 99% min. (GC)
Linear Formula (CH3)2SiCl2
Solubility Information Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
Formula Weight 129.06
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Dichlorodimethylsilane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 200-901-