Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dichloromethylvinylsilane, 97%

Manufacturer:  Alfa Aesar™ B2311909

Quantity 10g

 View more versions of this product

Catalog No. AAB2311909


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

124-70-9
141.07
UN2985
Methylvinyldichlorosilane; Vinylmethyldichlorosilane
C3H6Cl2Si
MFCD00000492
1740822
Dichloromethylvinylsilane

Specifications

124-70-9
141.07
UN2985
Methylvinyldichlorosilane; Vinylmethyldichlorosilane
92° to 93°C
10g
Dichloromethylvinylsilane
C3H6Cl2Si
MFCD00000492
1740822
Acrid
−78°C
4°C (39°F)
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

RTECSNumber : VV3533000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit