Alfa Aesar™ Dichloromethylvinylsilane, 97%

Manufacturer: Alfa Aesar™ B2311922

Quantity 100g

 View more versions of this product

Catalog No. AAB2311922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124-70-9
    Molecular Formula C3H6Cl2Si
    Formula Weight 141.07
    MDL Number MFCD00000492
    UN Number UN2985
    Beilstein 1740822
    Synonym Methylvinyldichlorosilane; Vinylmethyldichlorosilane
    Chemical Name or Material Dichloromethylvinylsilane

    Specifications

    Formula Weight 141.07
    TSCA TSCA
    Odor Acrid
    MDL Number MFCD00000492
    UN Number UN2985
    Boiling Point 92° to 93°C
    Melting Point −78°C
    Quantity 100g
    Flash Point 4°C (39°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
    RTECS Number VV3533000
    Molecular Formula C3H6Cl2Si
    CAS 124-70-9
    Beilstein 1740822
    Synonym Methylvinyldichlorosilane; Vinylmethyldichlorosilane
    Chemical Name or Material Dichloromethylvinylsilane
    Assay 97%

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit