Alfa Aesar™ Dichlorophenylborane, 96%

Manufacturer: Alfa Aesar™ 3258314

Quantity 25g

 View more versions of this product

Catalog No. AA3258314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 873-51-8
    Formula Weight 158.83
    MDL Number MFCD00013612
    UN Number UN2924
    Beilstein 2243576
    Synonym Phenylboron dichloride
    Chemical Name or Material Dichlorophenylborane

    Specifications

    Formula Weight 158.83
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Acrid
    CAS 873-51-8
    Synonym Phenylboron dichloride
    Density 1.194g/mL
    Sensitivity Moisture Sensitive
    Physical Form Liquid
    Beilstein 2243576
    Health Hazard 1 Danger
    Boiling Point 175°C
    Melting Point 7°C
    MDL Number MFCD00013612
    Refractive Index 1.545
    UN Number UN2924
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Flash Point 14° (57°F)
    Quantity 25g
    Color Colorless
    Chemical Name or Material Dichlorophenylborane
    Assay 96%

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit