Alfa Aesar™ Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

Manufacturer: Alfa Aesar™ H27752MC

Quantity 100mg

 View more versions of this product

Catalog No. AAH27752MC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 92220-65-0
    Molecular Formula C44H32Cl2Ir2N4
    Formula Weight 1072.09
    MDL Number MFCD07784515
    Synonym [(ppy)2IrCl]2
    Chemical Name or Material Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

    Specifications

    Formula Weight 1072.09
    Molecular Formula C44H32Cl2Ir2N4
    MDL Number MFCD07784515
    CAS 92220-65-0
    Synonym [(ppy)2IrCl]2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 100mg
    Chemical Name or Material Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit