Alfa Aesar™ Dichlorotris(triphenylphosphine)ruthenium(II), Premion™, 99.95% (metals basis), Ru 10.2% min

Manufacturer: Alfa Aesar™ 1050403

Quantity 1g

 View more versions of this product

Catalog No. AA1050403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15529-49-4
    Formula Weight 958.86
    MDL Number MFCD00013077
    Synonym Tris(triphenylphosphine)ruthenium(II) chloride
    Chemical Name or Material Dichlorotris(triphenylphosphine)ruthenium(II)
    Grade Premion™

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 958.86
    Color Brown to black
    Synonym Tris(triphenylphosphine)ruthenium(II) chloride
    Solubility Information Very slightly soluble (with dissociation) in acetone, alcohol, chloroform, ethyl acetate, and toluene
    Concentration Ru 10.2% min.
    Sensitivity Air & Moisture Sensitive
    Quantity 1g
    MDL Number MFCD00013077
    CAS 15529-49-4
    Melting Point 132° to 134°C
    Chemical Name or Material Dichlorotris(triphenylphosphine)ruthenium(II)
    Assay 99.95%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit