Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dickson™ Microprocessor-Based Temperature/Humidity/Dew Point Recorder Replacement Parts: Charts

Circular, paper charts for use with Dickson Temperature/Humidity/Dew Point Recorder.

Manufacturer:  Dickson™ C481

 View more versions of this product

Catalog No. 13-940-1008


  Check Availability
Add to cart

Specifications

Description

  • Charts measure 20.3cm in diameter
  • Running times available: 1, 7, or 31 days
  • Come in boxes of 60
Description & Specifications

Specifications

Dickson microprocessor-based temperature/humidity/dew point recorder
8 in.
Paper
31 Days
4.44° to 43.33°C
Circular Chart
20.3cm
60 Pk.
40° to 110°F
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit