Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dickson™ Replacement Charts for Super-Compact SC3 Temperature Chart Recorders
For SC3 Temperature Recorders.
Supplier: Dickson™ C320
Description
- Seven-day, 3 in. diameter (76mm) charts
- Comes in package of 60 sheets
Specifications
3 in. | |
7.6cm | |
For super-compact SC3 temperature chart |
Fahrenheit Chart | |
22°F to 68°F | |
-5° to 20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction