Learn More
AdipoGenSupplier Diversity Partner DICLOFENAC ACID(25G) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner CDXD0147G025
Diclofenac acid, AdipoGen Life Sciences. Chemical. CAS: 15307-86-5. Formula: C14H11Cl2NO2. MW: 296.15. Synthetic Nonsteroidal anti-inflammatory drug (NSAID) taken or applied to reduce inflammation and as an analgesic reducing pain. The primary mechanism responsible for its anti-inflammatory, antipyretic, and analgesic action is thought to be inhibition of prostaglandin synthesis by inhibition of cyclooxygenase (COX). It also appears to exhibit bacteriostatic activity by inhibiting bacterial DNA synthesis. Can be used as a reference compound.
Catalog No. 50-114-7011